3-Amino-7-bromoisoquinoline ≥95%

Supplier: Matrix Scientific

072592-250MG 072592-1G
102525-688EA 967.69 USD
102525-688 102525-690
3-Amino-7-bromoisoquinoline ≥95%
3-Amino-7-bromoisoquinoline

Hazard: Irritant

Formula: C₉H₇BrN₂
MDL Number: MFCD11040207
CAS Number: 1192815-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR