2-Amino-5-bromo-3-methylpyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-methylpyrazin-2-amine

072590-250MG 072590-1G
102525-678EA 826.15 USD
102525-678 102525-680
2-Amino-5-bromo-3-methylpyrazine ≥95%
2-Amino-5-bromo-3-methylpyrazine

Hazard: Irritant

Formula: C₅H₆BrN₃
MW: 188.0300 g/mol
MDL Number: MFCD08705773
CAS Number: 74290-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR