1-Boc-4-(6-chloro-5-nitro-4-pyrimidinyl)piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-4-(6-Chloro-5-nitro-4-pyrimidinyl)piperazine

072589-250MG 072589-1G 072589-5G
102525-672EA 960 USD
102525-672 102525-674 102525-676
1-Boc-4-(6-chloro-5-nitro-4-pyrimidinyl)piperazine ≥95%
1-Boc-4-(6-chloro-5-nitro-4-pyrimidinyl)piperazine

Hazard: Irritant

Formula: C₁₃H₁₈ClN₅O₄
MW: 343.7700 g/mol
MDL Number: MFCD11040204
CAS Number: 147539-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR