6-Chloro-N-methyl-5-nitro-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

072587-250MG 072587-1G 072587-5G
102525-660EA 355.38 USD
102525-660 102525-662 102525-664
6-Chloro-N-methyl-5-nitro-4-pyrimidinamine ≥95%
6-Chloro-N-methyl-5-nitro-4-pyrimidinamine

Hazard: Irritant

Formula: C₅H₅ClN₄O₂
CAS Number: 23126-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR