6-Chloro-N4-propylpyrimidine-4,5-diamine ≥95%

Supplier: Matrix Scientific

072586-250MG 072586-1G 072586-5G
102525-654EA 498.46 USD
102525-654 102525-656 102525-658
6-Chloro-N4-propylpyrimidine-4,5-diamine ≥95%
6-Chloro-N4-propylpyrimidine-4,5-diamine

Hazard: Irritant

Formula: C₇H₁₁ClN₄
MDL Number: MFCD07440144
CAS Number: 195252-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR