6-Ethoxypyridazin-3-amine ≥95%

Supplier: Matrix Scientific

072583-1G 072583-5G 072583-10G
102525-636EA 427.69 USD
102525-636 102525-638 102525-640
6-Ethoxypyridazin-3-amine ≥95%
6-Ethoxypyridazin-3-amine

Hazard: Irritant

Formula: C₆H₉N₃O
MW: 139.1552 g/mol
MDL Number: MFCD08460377
CAS Number: 39614-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR