5-Amino-2,6-dichloroquinoline ≥95%

Supplier: Matrix Scientific

072581-250MG 072581-1G
102525-626EA 1024.62 USD
102525-626 102525-628
5-Amino-2,6-dichloroquinoline ≥95%
5-Amino-2,6-dichloroquinoline

Hazard: Irritant

Formula: C₉H₆Cl₂N₂
MDL Number: MFCD09835367
CAS Number: 607380-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR