3-Bromoimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

072580-1G 072580-5G
102525-622EA 141.54 USD
102525-622 102525-624
3-Bromoimidazo[1,2-a]pyrazine ≥95%
3-Bromoimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C6H4BrN3
MW: 198.0200 g/mol
MDL Number: MFCD06245372
CAS Number: 57948-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR