2-Chloro-4-hydrazinopyridine ≥95%

Supplier: Matrix Scientific

072578-1G 072578-5G
102525-612EA 123.08 USD
102525-612 102525-614
2-Chloro-4-hydrazinopyridine ≥95%
2-Chloro-4-hydrazinopyridine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD08460247
CAS Number: 700811-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR