6-Bromo-4-chloropyrido[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

072575-250MG 072575-1G
102525-598EA 1776.92 USD
102525-598 102525-600
6-Bromo-4-chloropyrido[2,3-d]pyrimidine ≥95%
6-Bromo-4-chloropyrido[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₇H₃BrClN₃
MW: 244.4758 g/mol
MDL Number: MFCD11040200
CAS Number: 1215787-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR