2,4-Dichloro-5-nitropyridine ≥95%

Supplier: Matrix Scientific

072573-1G 072573-5G
102525-590EA 36.92 USD
102525-590 102525-592
2,4-Dichloro-5-nitropyridine ≥95%
2,4-Dichloro-5-nitropyridine

Hazard: Irritant

Formula: C₅H₂Cl₂N₂O₂
MW: 192.9900 g/mol
MDL Number: MFCD07368834
CAS Number: 4487-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR