Methyl-1-(2-chloropyrimidin-4-yl)piperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

072571-250MG 072571-1G 072571-5G
102525-576EA 640 USD
102525-576 102525-578 102525-580
Methyl-1-(2-chloropyrimidin-4-yl)piperidine-4-carboxylate ≥95%
Methyl-1-(2-chloropyrimidin-4-yl)piperidine-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₄ClN₃O₂
CAS Number: 889126-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR