4-Bromo-3-nitrobenzene-1,2-diamine ≥95%

Supplier: Matrix Scientific

072570-250MG 072570-1G 072570-5G
102525-570EA 456.92 USD
102525-570 102525-572 102525-574
4-Bromo-3-nitrobenzene-1,2-diamine ≥95%
4-Bromo-3-nitrobenzene-1,2-diamine

Hazard: Irritant

Formula: C₆H₆BrN₃O₂
MW: 232.0400 g/mol
MDL Number: MFCD11040197
CAS Number: 147021-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR