4-Bromo-3-nitrophenol ≥95%

Supplier: Matrix Scientific

072569-1G 072569-5G
102525-564EA 44.62 USD
102525-564 102525-566
4-Bromo-3-nitrophenol ≥95%
4-Bromo-3-nitrophenol

Hazard: Irritant

Formula: C₆H₄BrNO₃
MDL Number: MFCD11040194
CAS Number: 78137-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR