2-Chloro-1-methoxy-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

072568-250MG 072568-1G 072568-5G
102525-558EA 355.38 USD
102525-558 102525-560 102525-562
2-Chloro-1-methoxy-3-nitrobenzene ≥95%
2-Chloro-1-methoxy-3-nitrobenzene

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD11035883
CAS Number: 3970-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR