2-Chloro-3-nitrophenol ≥95%

Supplier: Matrix Scientific

072567-1G 072567-5G 072567-10G
102525-552EA 533.85 USD
102525-552 102525-554 102525-556
2-Chloro-3-nitrophenol ≥95%
2-Chloro-3-nitrophenol

Hazard: Irritant

Formula: C₆H₄ClNO₃
MDL Number: MFCD11035882
CAS Number: 603-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR