3,6-Dimethylsalicylaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-3,6-dimethylbenzaldehyde, 3,6-Dimethyl-2-hydroxybenzaldehyde

072565-250MG 072565-1G 072565-5G
102525-540EA 400 USD
102525-540 102525-542 102525-544
3,6-Dimethylsalicylaldehyde ≥95%
3,6-Dimethylsalicylaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.1700 g/mol
MDL Number: MFCD04037328
CAS Number: 1666-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR