5-Bromo-6-methylpyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-5-bromo-6-methylpyrimidine

072562-250MG 072562-1G
102525-526EA 967.69 USD
102525-526 102525-528
5-Bromo-6-methylpyrimidin-4-amine ≥95%
5-Bromo-6-methylpyrimidin-4-amine

Hazard: Irritant

Formula: C₅H₆BrN₃
MW: 188.03 g/mol
MDL Number: MFCD00626351
CAS Number: 7752-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR