6-Methylpyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

072561-250MG 072561-1G
102525-522EA 283.68 USD
102525-522 102525-524
6-Methylpyrimidin-4-amine ≥95%
6-Methylpyrimidin-4-amine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.13 g/mol
Melting Pt: 197 °C
MDL Number: MFCD01863677
CAS Number: 3435-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR