2-Amino-3-bromo-5-fluoropyridine ≥95%

Supplier: Matrix Scientific

072560-1G 072560-5G 072560-10G
102525-516EA 178.46 USD
102525-516 102525-518 102525-520
2-Amino-3-bromo-5-fluoropyridine ≥95%
2-Amino-3-bromo-5-fluoropyridine

Hazard: Irritant

Formula: C₅H₄BrFN₂
MDL Number: MFCD03092924
CAS Number: 869557-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR