8-Bromoquinazolin-4(1H)-one ≥95%

Supplier: Matrix Scientific

072558-250MG 072558-1G 072558-5G
102525-504EA 363.54 USD
102525-504 102525-506 102525-508
8-Bromoquinazolin-4(1H)-one ≥95%
8-Bromoquinazolin-4(1H)-one

Hazard: Irritant

Formula: C₈H₅BrN₂O
MW: 225.0421 g/mol
MDL Number: MFCD10699396
CAS Number: 77150-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR