2-Chloro-N-4-methylpyridine-3,4-diamine ≥95%

Supplier: Matrix Scientific

072557-250MG 072557-1G 072557-5G
102525-498EA 355.38 USD
102525-498 102525-500 102525-502
2-Chloro-N-4-methylpyridine-3,4-diamine ≥95%
2-Chloro-N-4-methylpyridine-3,4-diamine

Hazard: Irritant

Formula: C₆H₈ClN₃
MDL Number: MFCD00778574
CAS Number: 50432-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR