Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloro-1H-1,2,4-triazole-3-carboxylate

072555-250MG 072555-1G 072555-5G
102525-486EA 427.69 USD
102525-486 102525-488 102525-490
Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate ≥95%
Ethyl-5-chloro-1H-1,2,4-triazole-3-carboxylate

Hazard: Irritant

Formula: C₅H₆ClN₃O₂
MW: 175.5700 g/mol
MDL Number: MFCD11040186
CAS Number: 774608-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR