5-Bromo-1H-1,2,4-triazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

072553-250MG 072553-1G 072553-5G
102525-474EA 427.69 USD
102525-474 102525-476 102525-478
5-Bromo-1H-1,2,4-triazole-3-carboxylic acid ≥95%
5-Bromo-1H-1,2,4-triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₃H₂BrN₃O₂
MW: 191.97 g/mol
MDL Number: MFCD11040182
CAS Number: 674287-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR