8-Bromo-4-chloroquinazoline ≥95%
Supplier: Matrix Scientific
072551-250MG
072551-1G
072551-5G
102525-462EA
569.23
USD
102525-462
102525-464
102525-466
8-Bromo-4-chloroquinazoline ≥95%
8-Bromo-4-chloroquinazoline
Hazard: Irritant
Formula:
C₈H₄BrClN₂ MW: 243.4900 g/mol |
MDL Number:
MFCD09738589 CAS Number: 125096-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...