4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine ≥95%

Supplier: Matrix Scientific

072550-250MG 072550-1G 072550-5G
102525-456EA 58.46 USD
102525-456 102525-458 102525-460
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine ≥95%
4-Chloro-1-methyl-1H-imidazo[4,5-c]pyridine

Hazard: Irritant

Formula: C₇H₆ClN₃
MW: 167.6000 g/mol
MDL Number: MFCD00222101
CAS Number: 50432-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR