Ethyl-7-bromo-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 7-bromo-1H-indole-2-carboxylate

072549-1G 072549-5G
102525-452EA 60 USD
102525-452 102525-454
Ethyl-7-bromo-1H-indole-2-carboxylate ≥95%
Ethyl-7-bromo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀BrNO₂
MW: 268.1066 g/mol
MDL Number: MFCD04972046
CAS Number: 16732-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR