4-(Pyrrolidin-1-yl)benzonitrile ≥95%
Supplier: Matrix Scientific
072548-1G
072548-5G
072548-10G
102525-444EA
178.46
USD
102525-444
102525-446
102525-448
4-(Pyrrolidin-1-yl)benzonitrile ≥95%
4-(Pyrrolidin-1-yl)benzonitrile
Hazard: Irritant
Formula:
C₁₁H₁₂N₂ MW: 172.23 g/mol |
MDL Number:
MFCD07368512 CAS Number: 10282-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...