2-Chloro-5-nitropyridin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-chloro-5-nitropyridine

072547-1G 072547-5G
102525-440EA 432.31 USD
102525-440 102525-442
2-Chloro-5-nitropyridin-4-amine ≥95%
2-Chloro-5-nitropyridin-4-amine

Hazard: Irritant

Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
Melting Pt: 189…191 °C
MDL Number: MFCD09033797
CAS Number: 2604-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR