2-Chloro-3-nitropyridin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-chloro-3-nitropyridine

072546-250MG 072546-1G 072546-5G
102525-432EA 15.38 USD
102525-432 102525-434 102525-436
2-Chloro-3-nitropyridin-4-amine ≥95%
2-Chloro-3-nitropyridin-4-amine

Hazard: Irritant

Formula: C₅H₄ClN₃O₂
MW: 173.5572 g/mol
MDL Number: MFCD07368898
CAS Number: 2789-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR