6-Bromoisoquinolin-1(2H)-one ≥95%

Supplier: Matrix Scientific

072545-1G 072545-5G
102525-428EA 67.69 USD
102525-428 102525-430
6-Bromoisoquinolin-1(2H)-one ≥95%
6-Bromoisoquinolin-1(2H)-one

Hazard: Irritant

Formula: C₉H₆BrNO
MDL Number: MFCD07374394
CAS Number: 82827-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR