Methyl-5-hydroxypyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

072544-1G 072544-5G 072544-10G
102525-420EA 61.54 USD
102525-420 102525-422 102525-424
Methyl-5-hydroxypyrazine-2-carboxylate ≥95%
Methyl-5-hydroxypyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₆N₂O₃
MDL Number: MFCD08273469
CAS Number: 13924-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR