tert-Butyl-4-(3-aminopyrazin-2-yl)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(3-aminopyrazin-2-yl)piperazine-1-carboxylate

072543-250MG 072543-1G 072543-5G
102525-414EA 498.46 USD
102525-414 102525-416 102525-418
tert-Butyl-4-(3-aminopyrazin-2-yl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(3-aminopyrazin-2-yl)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₁N₅O₂
MDL Number: MFCD11040180
CAS Number: 1208542-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR