3-Iodo-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Iodoindazole

072541-1G 072541-5G 072541-25G
102525-402EA 44.62 USD
102525-402 102525-404 102663-874
3-Iodo-1H-indazole ≥95%
3-Iodo-1H-indazole

Hazard: Irritant

Formula: C₇H₅IN₂
MW: 244.0325 g/mol
MDL Number: MFCD04972469
CAS Number: 66607-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR