4,6-Diiodo-2-methylpyrimidine ≥95%

Supplier: Matrix Scientific

072539-250MG 072539-1G 072539-5G
102525-390EA 427.69 USD
102525-390 102525-392 102525-394
4,6-Diiodo-2-methylpyrimidine ≥95%
4,6-Diiodo-2-methylpyrimidine

Hazard: Irritant

Formula: C₅H₄I₂N₂
CAS Number: 66298-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR