Methyl-5-bromopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromopyrazine-2-carboxylate

072537-1G 072537-5G
102525-380EA 261.54 USD
102525-380 102525-382
Methyl-5-bromopyrazine-2-carboxylate ≥95%
Methyl 5-bromopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD09991715
CAS Number: 210037-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR