1H-Imidazo[4,5-b]pyridin-2(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Dihydro-2H-imidazo[4,5-b]pyridin-2-one

072536-1G 072536-5G
102525-374EA 710.77 USD
102525-374 102525-376
1H-Imidazo[4,5-b]pyridin-2(3H)-one ≥95%
1H-Imidazo[4,5-b]pyridin-2(3H)-one

Hazard: Irritant

Formula: C₆H₅N₃O
MW: 135.1200 g/mol
MDL Number: MFCD00464845
CAS Number: 16328-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR