7-Fluoro-1-tetralone ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Fluoro-3,4-dihydronaphthalen-1(2H)-one

072530-1G 072530-5G
102525-342EA 84.62 USD
102525-342 102525-344
7-Fluoro-1-tetralone ≥95%
7-Fluoro-1-tetralone

Hazard: Irritant

Formula: C₁₀H₉FO
MW: 164.18 g/mol
MDL Number: MFCD07375433
CAS Number: 2840-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR