1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
7-Chloro-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid
072529-1G
072529-5G
072529-25G
102525-334EA
100
USD
102525-334
102525-336
102551-794
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid ≥95%
1-Cyclopropyl-6-fluoro-7-chloro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₈ClFN₂O₃ MW: 282.6600 g/mol |
MDL Number:
MFCD08458865 CAS Number: 100361-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...