2,6-Difluoro-4-hydroxybenzyl alcohol ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Difluoro-4-(hydroxymethyl)phenol

072524-250MG 072524-1G 072524-5G
102525-304EA 150.77 USD
102525-304 102525-306 102525-308
2,6-Difluoro-4-hydroxybenzyl alcohol ≥95%
2,6-Difluoro-4-hydroxybenzyl alcohol

Hazard: Irritant

Formula: C₇H₆F₂O₂
MW: 160.1200 g/mol
MDL Number: MFCD11040173
CAS Number: 438049-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR