2-Chlorobenzothiazole-6-carbonitrile ≥95%

Supplier: Matrix Scientific

072523-250MG 072523-1G 072523-5G
102525-298EA 640 USD
102525-298 102525-300 102525-302
2-Chlorobenzothiazole-6-carbonitrile ≥95%
2-Chlorobenzothiazole-6-carbonitrile

Hazard: Irritant

Formula: C₈H₃ClN₂S
MW: 194.6408 g/mol
MDL Number: MFCD09743933
CAS Number: 80945-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR