3-Nitro-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

072521-250MG 072521-1G 072521-5G
102525-286EA 498.46 USD
102525-286 102525-288 102525-290
3-Nitro-1H-pyrazole-4-carbonitrile ≥95%
3-Nitro-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C4H2N4O2
MW: 138.0800 g/mol
MDL Number: MFCD00195039
CAS Number: 39205-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR