(5-Methylpyrazin-2-yl)methanol ≥95%

Supplier: Matrix Scientific

072516-250MG 072516-1G 072516-5G
102525-256EA 355.38 USD
102525-256 102525-258 102525-260
(5-Methylpyrazin-2-yl)methanol ≥95%
(5-Methylpyrazin-2-yl)methanol

Hazard: Irritant

Formula: C₆H₈N₂O
MW: 124.1405 g/mol
MDL Number: MFCD11044903
CAS Number: 61892-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR