tert-Butyl (5-methylpyrazin-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Boc-amino)-5-methylpyrazine

072515-1G 072515-5G 072515-10G
102525-250EA 92.31 USD
102525-250 102525-252 102525-254
tert-Butyl (5-methylpyrazin-2-yl)carbamate ≥95%
tert-Butyl (5-methylpyrazin-2-yl)carbamate

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MW: 209.2500 g/mol
MDL Number: MFCD09907674
CAS Number: 369638-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR