5-tert-Butyl-1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 5-tert-Butyl 1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate

072514-250MG 072514-1G 072514-5G
102525-244EA 684.62 USD
102525-244 102525-246 102525-248
5-tert-Butyl-1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate ≥95%
5-tert-Butyl-1-ethyl 3-aminopyrrolo[3,4-c]pyrazole-1,5(4H,6H)-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₂₀N₄O₄
MW: 296.3223 g/mol
MDL Number: MFCD08061258
CAS Number: 398495-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR