Imidazo[1,2-b]pyridazin-6-amine ≥95%

Supplier: Matrix Scientific

072513-250MG 072513-1G 072513-5G
102525-238EA 640 USD
102525-238 102525-240 102525-242
Imidazo[1,2-b]pyridazin-6-amine ≥95%
Imidazo[1,2-b]pyridazin-6-amine

Hazard: Irritant

Formula: C₆H₆N₄
MDL Number: MFCD10698052
CAS Number: 6653-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR