6-Chloroimidazo[1,2-b]pyridazin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-6-chloroimidazo[1,2-b]pyridazine

072512-1G 072512-5G
102525-234EA 253.85 USD
102525-234 102525-236
6-Chloroimidazo[1,2-b]pyridazin-3-amine ≥95%
6-Chloroimidazo[1,2-b]pyridazin-3-amine

Hazard: Irritant

Formula: C₆H₅ClN₄
MW: 168.5800 g/mol
MDL Number: MFCD09881451
CAS Number: 166176-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR