4-Chlorobenzo[d]thiazole ≥95%

Supplier: Matrix Scientific

072510-1G 072510-5G
102525-222EA 710.77 USD
102525-222 102525-224
4-Chlorobenzo[d]thiazole ≥95%
4-Chlorobenzo[d]thiazole

Hazard: Irritant

Formula: C₇H₄ClNS
MW: 169.6300 g/mol
MDL Number: MFCD11040158
CAS Number: 3048-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR