tert-Butyl-3-cyano-4-hydroxypyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

072509-1G 072509-5G
102525-218EA 355.38 USD
102525-218 102525-220
tert-Butyl-3-cyano-4-hydroxypyrrolidine-1-carboxylate ≥95%
tert-Butyl-3-cyano-4-hydroxypyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₃
CAS Number: 197143-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR