tert-Butyl (3-(hydroxymethyl)pyridin-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-3-(hydroxymethyl)pyridine, 2-BOC protected

072508-1G 072508-5G 072508-10G
102525-212EA 512.31 USD
102525-212 102525-214 102525-216
tert-Butyl (3-(hydroxymethyl)pyridin-2-yl)carbamate ≥95%
tert-Butyl (3-(hydroxymethyl)pyridin-2-yl)carbamate

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₃
CAS Number: 877593-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR